n-Hexane ≥95%, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

HX0291-1 HX0291P-1
CAHX0291-1CS 2076 CAD
CAHX0291-1 CAHX0291P-1 CAHX0291P-6
n-Hexane ≥95%, Supelco®
n-Hexane

HPLC, Meets Reagent Specifications for testing USP/NF monographs

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69 °C (1013 hPa)
Melting Pt: –94.3 °C
Density: 0.659 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid, free from particulates
Color (APHA) 10 max
Residue after evaporation 5ppm max
Infrared spectrum Conforms
Assay (n-hexane) 95% min
Sulfur compounds (as S) 0.005% max
Thiophene To pass test
Water-soluble titrable acid 0.0003meq/g max
Assay (total hexane isomers + methylcyclopentane) 99.0% min
UV Abs. at 200nm 1.0AU max
UV Abs. at 210 nm 0.30AU max
UV Abs. at 220 nm 0.10AU max
UV Abs. at 230 nm 0.05AU max
UV Abs. at 240 nm 0.04AU max
UV Abs. at 250 nm 0.02AU max
UV Abs. at 270 nm 0.01AU max
UV Abs. at 300 nm 0.01AU max
UV Abs. at 350 nm 0.01AU max
UV Cut-off 200nm max
Water (H2O) 0.01% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR