Media Bottles, PET, Sterile, Greiner Bio-One
Supplier: Greiner Bio-One
950700 951700 951700P 952700
82051-492CS 306.8 CAD
82051-492 82051-494 82051-592 82051-594
Media Bottles, PET, Sterile, Greiner Bio-One
Bottles Cell Culture Bottles
These sterile PET media bottles are ideal for storage of media, buffers, and laboratory samples.
  • Clear polyethylene terephthalate construction
  • Sturdy and crack-resistant
  • Flexible
  • Free of RNase, DNase, and human DNA
  • Nonpyrogenic and noncytotoxic
The square design of the media bottles allows efficient use of storage space.
Clear polyethylene terephthalate (PET) construction makes content identification simple. The sturdy yet flexible crack-resistant bottles are supplied with white screw caps. All bottles are graduated to the working volume and supplied sterile.