STERLING™ Nitrile and STERLING™ Nitrile-XTRA™ Exam Gloves, Kimberly-Clark
Supplier: Microflex
50705 50706 50707 50708 50709 53137 53138 53139 53140 53141
CA89140-816CS 517.35 CAD
CA89140-816 CA89140-818 CA89140-820 CA89140-822 CA89140-824 CA94023-906 CA94023-908 CA94023-910 CA94023-912 CA94023-914
STERLING™ Nitrile and STERLING™ Nitrile-XTRA™ Exam Gloves, Kimberly-Clark
Gloves
Powder-free, latex-free gloves provide the protection of nitrile with sensitivity comparable to natural rubber latex.
  • Excellent tactile sensitivity allows easy handling of delicate instruments
  • Ambidextrous, textured fingertips, beaded cuff
  • Powder-free
  • Thickness: 3.5 mil (finger), 3.1 mil (palm), 2.8 mil (cuff)
Excellent tactile sensitivity allows easy handling of delicate instruments. Ambidextrous. Beaded cuff. Textured fingertips.
Certifications: Gloves are tested with chemotherapy drugs according to ASTM D6978.
Ordering information: For the 24.More Product Information