n-Heptane ≥99.0% (by GC), OmniSolv®, Extra Low UV Cut-off, Supelco®

Supplier: MilliporeSigma

Synonyms: high purity solvents

HX0076-1 HX0076-1
CAHX0076-1EA 523.53 CAD
CAHX0076-1 EM-HX0076-1
n-Heptane ≥99.0% (by GC), OmniSolv®, Extra Low UV Cut-off, Supelco®
n-Heptane

For critical measurements in spectrophotometry and liquid chromatography. Low UV cutoff 193nm. Certificate of Analysis on label.

Formula: H₃C(CH₂)₅CH₃
MW: 100.20 g/mol
Boiling Pt: 98.4 °C (1013 hPa)
Melting Pt: –90.6 °C
Density: 0.6838 g/cm³ (20 °C)
Flash Pt: –4 °C
MDL Number: MFCD00009544
CAS Number: 142-82-5
UN: 1206
ADR: 3,II
Merck Index: 13,04679
https://ca.vwr.com/stibo/web/std.lang.all/64/15/736415.jpg

Order Now

Specification Test Results

Assay (n−heptane) min. 99.0%
Assay (saturated C7 hydrocarbons) min 99.9%
Color (APHA) max 10
Evaporation residue max 1 ppm
Filtered through 0.2 μm filter Passes test
Fluorescence (as quinine base) max 250 ppt
Form Clear liquid
Infrared Spectrum Conforms
UV Abs. at 193 nm max 1.00 AU
UV Abs. at 200 nm max 0.80 AU
UV Abs. at 210 nm max 0.30 AU
UV Abs. at 220 nm max 0.15 AU
UV Abs. at 260 nm max 0.01 AU
UV Cut−off max 193 nm
Water (H2O) max 0.02 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR