Alcohol Tralle and Proof Hydrometers

Supplier: Heathrow Scientific
6612-1 6612-2TS
76546-478EA 44.77 CAD
76546-478 76546-480
Alcohol Tralle and Proof Hydrometers
Hydrometers
Our alcohol hydrometers measure using the tralle and proof scales.

  • SafetyBLUE fill
  • Thermometer (model 6612-2TS only) measures from −10 to +100 °F
  • Tralle/proof range 0 to 100%/0 to 200
  • Tralle/proof subdivisions/tolerance 1%/ ±1%, 2%/ ±2%
  • Calibration for use at 15.5 °C (60 °F)
  • Not for use with beer or wine

Verify the alcohol content of distilled spirits with our alcohol hydrometer with Tralle and proof scales. Model 6612-2TS includes an internal thermometer for measuring solution temperature.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR