HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid free acid ≥99.0%, OmniPur® for biotechnology, Millipore®

Supplier: MilliporeSigma

Synonyms: HEPES free acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulfonic acid, 4-(2-Hydroxyethyl)piperazin-1-ylethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid), 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid

5310-100GM 5320-500GM 5340-5KG 5330-1KG 5330-1KG
CA-EM5310EA 229.86 CAD
CA-EM5310 CA-EM5320 CA-EM5340 CAEM5330 CAEM5330-1KG
HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid free acid ≥99.0%, OmniPur® for biotechnology, Millipore®
HEPES (4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid)

Free acid for molecular biology. DNase-, RNase-, protease-, and phosphatase-free.

Formula: C₈H₁₈N₂O₄S
MW: 238.31 g/mol
Melting Pt: 210…215 °C
Density: 1.448 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006158
CAS Number: 7365-45-9
EINECS: 230-907-9
Merck Index: 13,04672
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cdc09c866676149941b93b2cac8d453175059a2&vid=0b09e5b84ea34a806443c3071fad63e15e4e3433&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR