2,2,4-Trimethylpentane ≥99% ACS, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: IsobutyltrimethylmethaneIsooctaneIsobutyltrimethylethane

032V-07
BDH1155-19LEA 1572.75 CAD
BDH1155-19L
2,2,4-Trimethylpentane ≥99% ACS, VWR Chemicals BDH®
2,2,4-Trimethylpentane

Clear, colorless liquid. Meets ACS specifications for general use.

Formula: (CH₃)₂CHCH₂C(CH₃)₃
MW: 114.23 g/mol
Boiling Pt: 99.2 °C (1013 hPa)
Melting Pt: –107 °C
Density: 0.692 g/cm³ (20 °C)
Flash Pt: –12 °C
MDL Number: MFCD00008943
CAS Number: 540-84-1
UN: 1262
ADR: 3,II
Merck Index: 13,05212
https://ca.vwr.com/stibo/web/std.lang.all/00/81/740081.jpg

Order Now

Specification Test Results

Appearance Clear, colorless liquid
Water (K.F.) Less than 0.010%
Residue Less than 0.001% (0.0010 g/145 mL)
GC Purity Greater than 99.0%
Color (APHA) Less than 10
Titrable acid Less than 0.3µeq/g (No more than 1.0mL of 0.01N sodium hydroxide)
Sulfur compounds Less than 0.005% as S

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR