Toluene ≥99.5% ACS, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Methylbenzene, Phenylmethane, FM approval

BDH1151-19L BDH1151-204L BDH1151-4LG
BDH1151-19LEA 486.67 CAD
BDH1151-19L BDH1151-204L BDH1151-4LG
Toluene ≥99.5% ACS, VWR Chemicals BDH®
Toluene

Suitable for histology and cytology applications.

Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 110.6
Melting Pt: -95
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 4
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3 II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless liquid
Water (K.F.) Less than 0.030%
Residue Less than 0.001% (0.0010 g/115 mL)
GC Purity Greater than 99.5%
Color (APHA) Less than 10
Substances darkened by sulfuric acid Passes test (Toluene layer should be colorless. The acid should be no darker than the standard)
Sulfur compounds (as S) Less than 0.003%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR