n-Heptane ≥99.0% ACS analytical reagent, general laboratory use
Supplier: HONEYWELL BURDICK & JACKSON CA
|
Danger
|
Synonyms:
high purity solvents
LP206-19
LP206-4
LP206-204
LP206-4P
CABJLP206D19EA
1420.01
CAD
CABJLP206D19
CABJLP206-4
BJLP206-204
BJLP206-4P
n-Heptane ≥99.0% ACS analytical reagent, general laboratory use
n-Heptane
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Formula:
H₃C(CH₂)₅CH₃ MW: 100.20 g/mol Boiling Pt: 98.4 Melting Pt: -90.6 Density: 0.684 g/cm³ (20 °C) Flash Pt: –4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009544 CAS Number: 142-82-5 EINECS: 205-563-8 UN: 1206 ADR: 3 II Merck Index: 13,04679 |
|
Specification Test Results
Appearance | Passes test |
Assay by GC Analysis (n-Heptane) | 99 % min. |
Assay by GC Analysis (total C7 isomers) | 99.9 % min. |
Color (APHA) | 10 max. |
Residue after Evaporation | 0.001 % max. |
Water by Karl Fischer Titration | 0.01 % max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...