Tin(IV) acetate ≥99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
89921-06
89921-14
CAAA89921-06EA
295.46
CAD
CAAA89921-06
CAAA89921-14
Tin(IV) acetate ≥99%
Tin (IV) acetate
Crystalline. Hygroscopic.
CAS No.: 2800-96-6
Molecular Formula: C8H12O8Sn
Formula Weight: 354.89
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: White
Odor: Light
Melting Point: 232-233°
MDL No.: MFCD00014976
Formula:
C₈H₁₂O₈Sn Melting Pt: 232…233 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014976 CAS Number: 2800-96-6 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...