Tellurium dioxide ≥99.9995% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning

10882-14 10882-22 10882-36
CAAA10882-14EA 237 CAD
CAAA10882-14 CAAA10882-22 CAAA10882-36
Tellurium dioxide ≥99.9995% (metals basis), Puratronic®
Tellurium dioxide

Powder. Purity based on metal contaminates only.


Purity: 99.9995% CAS No.: 7446-07-3 Molecular Formula: TeO2 Formula Weight: 159.60 Physical Form: Powder Density: 5.8 Melting Point: 733° Merck Reference: 14,9123 MDL No.: MFCD00011263

Formula: O₂Te
MW: 159.6 g/mol
Boiling Pt: 1245 °C (1013 hPa)
Melting Pt: 733 °C
Density: 5.8
Storage Temperature: Ambient
MDL Number: MFCD00011263
CAS Number: 7446-07-3
EINECS: 231-193-1
Merck Index: 14,9123

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR