Titanium ≥99% (metals basis), powder -325 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
10386-09
10386-22
10386-36
10386-D3
CAAA10386-09EA
29.7
CAD
CAAA10386-09
CAAA10386-22
CAAA10386-36
CAAA10386-D3
Titanium ≥99% (metals basis), powder -325 mesh
Titanium
MDL: MFCD00011264
Purity: 99%
CAS No.: 7440-32-6
Appearance: Grey
Odor: Odorless
UN No.: 2546
MDL No.: MFCD00011264
Formula:
Ti Boiling Pt: 3277 °C (1013 hPa) Melting Pt: 1677 °C Density: 4.5 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011264 CAS Number: 7440-32-6 EINECS: 231-142-3 UN: 2546 ADR: 4.2,II |
Specification Test Results
Assay (by Metals Basis) | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...