Hydrofluoric acid 48 - 51% ACS, VWR Chemicals BDH®

Supplier: VWR International
Danger

263000-80601 263000-80401 263000-80616
CABDH3040PCS 3224.82 CAD
CABDH3040P CABDH30423.8LP CABDH30422.5LP
Hydrofluoric acid 48 - 51% ACS, VWR Chemicals BDH®
Hydrofluoric acid
Formula: HF
MW: 20.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011346
CAS Number: 7664-39-3
EINECS: 231-634-8
UN: 1790
ADR: 8,II

Order Now

Specification Test Results

Assay (HF) 48.0 - 51.0%
Appearance Clear, colorless liquid
Arsenic (As) 0.05 ppm max.
Chloride (Cl) 5 ppm max.
Copper (Cu) 0.1 ppm max.
Fluosilicic acid (H2SiF6) 0.01% max.
Heavy metals (as Pb) 0.5 ppm max.
Iron (Fe) 1 ppm max.
Phosphate (PO4) 1 ppm max.
Residue after ignition 5 ppm max.
Sulfate and sulfite (as SO4) 5 ppm max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR