Hydrochloric acid 36.5 - 38.0% ACS, NF, FCC, VWR Chemicals BDH®
Supplier: VWR International
|
Danger
|
Synonyms:
Hydrochloric acid solution, Muriatic acid
257900-21601
257900-22001
257900-80601
257900-80602
257900-80603
257900-80401
257900-80606
CABDH303419LEA
3539.67
CAD
CABDH303419L
CABDH3038191L
CABDH3026P
CABDH30282.5LG
CABDH3030PC
CABDH30323.8LP
CABDH3128-2.2P
Hydrochloric acid 36.5 - 38.0% ACS, NF, FCC, VWR Chemicals BDH®
Hydrochloric acid
Free from particulates. Meets reagent specifications for testing USP/NF monographs.
Formula:
HCl MW: 36.46 g/mol Boiling Pt: 110 °C (1013 hPa) Melting Pt: –30 °C Density: 1.18 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8,II |
Specification Test Results
Assay (HCl) | 36.5-38.0% |
Assay (HCl) Be' | 22.7 deg Be' min. |
Ammonium (NH4) | 0.0003% max. |
Appearance | Clear, colorless fuming liquid |
Arsenic (As) | 10 ppb max. |
Bromide or Iodide | To pass test |
Clarity of Solution | Free from suspended matter/sediment |
Color (APHA) | 10 max. |
Free Bromine or Chlorine | To pass test |
Free Chlorine (Cl2) | 0.0001% max. |
Heavy Metals (as Pb) | 0.0001% max. |
Identification FCC | To pass test |
Identification NF | To pass test |
Iron (Fe) | 0.00002% max |
Lead (Pb) FCC/USP | 1 mg/kg max. |
Mercury FCC | 0.10 ppm max. |
Organic Compounds - Benzene | 0.05 ppm max. |
Organic Compounds - Total fluorinated organic compounds | 25 ppm max. |
Reducing Substances | To pass test |
Residue After Ignition | 0.0005% max. |
Specific Gravity at 60°F | Record only (approx. 1.17-1.20) |
Sulfate (SO4) | 0.0001% max. |
Sulfite (SO3) | 0.0001% max. |
Total Organic Compounds | 5 ppm max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...