Hexane (mixture of isomers) ACS, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Hexanes, Isohexane

BDHVBDH1129-4LG BDHVBDH1129-4LP BDHVBDH1129-19L BDHVBDH1129-204L
BDH1129-4LGCS 1113.64 CAD
BDH1129-4LG BDH1129-4LP BDH1129-19L BDH1129-204L
Hexane (mixture of isomers) ACS, VWR Chemicals BDH®
Hexane (mixture of isomers)

Meets ACS specifications for general use.

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 68…70 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.66 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (clear, colorless liquid) Passes Test
ACS - Assay (as n-hexane) (by GC) For Information Only
ACS - Assay (sum of 5 isomers, total hexanes, plus methylcyclopentane)(by GC) ≥ 98.5 %
ACS - Color (APHA) ≤ 10
ACS - Residue after Evaporation ≤ 0.001 %
ACS - Sulfur Compounds (as S) ≤ 0.005 %
ACS - Thiophene Passes Test
ACS - Water-Soluble Titrable Acid (meq/g) ≤ 0.0003

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR