n-Heptane ≥99%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
|
Danger
|
BDH1127-4LG
BDH1127-19L
BDH1127-204L
BDH1127-4LP
BDH1127-4LGCS
1965.6
CAD
BDH1127-4LG
BDH1127-19L
BDH1127-204L
BDH1127-4LP
n-Heptane ≥99%, Laboratory Reagent, VWR Chemicals BDH®
n-Heptane
Clear liquid.
Formula:
H₃C(CH₂)₅CH₃ Boiling Pt: 98.4 °C (1013 hPa) Melting Pt: –90.6 °C Density: 0.6838 g/cm³ (20 °C) Flash Pt: –4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009544 CAS Number: 142-82-5 EINECS: 205-563-8 UN: 1206 ADR: 3,II Merck Index: 13,04679 |
|
Specification Test Results
Appearance | Clear |
Water (K.F.) | Less than 0.010% |
Residue | Less than 0.001% (0.0010 g/146 mL) |
GC Purity | Greater than 99.9% n-heptane (n-C7) and C7 isomers (Typical - 99.9+%) |
n-Heptane | Greater than 99.0% |
Color (APHA) | Less than 10 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...