Falcon® Cell Culture Flasks, Corning
Supplier: Corning
355000 355001 353009 353014 353018 353024 353028 353107 353109 353110 353112 353082 353133 353135 353136 353138 353139 353108 353139 353082 353138
CA15705-072CS 423.54 CAD
CA15705-072 CA15705-074 CA29185-060 CA29185-078 CA29185-085 CA29185-087 CA29185-258 CA29185-298 CA29185-302 CA29185-304 CA29185-308 CABD353082 CABD353133 CABD353135 CABD353136 CABD353138 CABD353139 CA29185-300 47743-884 29185-053 47743-882
Falcon® Cell Culture Flasks, Corning
Flasks Cell Culture Flasks
PS, tissue culture-treated, sterile, non-pyrogenic.
  • Easy to read printed volumetric graduations, and writing patch
  • Vented caps incorporate a 0.2 µm hydrophobic membrane
  • Convenient, peel-open, medical-style packaging ensures flask sterility is maintained
Vacuum gas plasma tissue culture treatment ensures consistent cell attachment, spreading and growth.
Barcoded (robotics/automation compatible).
Caution: NOTE: Culture area and capacity are nominal.