Hexaammineruthenium trichloride Ru ≥ 32.1% Ru ≥ 32.1%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Hexaammineruthenium(III) chloride, Hexaammineruthenium (III) chloride

10511-01 10511-03 10511-06
CAAA10511-01EA 82.16 CAD
CAAA10511-01 CAAA10511-03 CAAA10511-06
Hexaammineruthenium trichloride Ru ≥ 32.1% Ru ≥ 32.1%
Hexaammineruthenium trichloride

CAS No.: 14282-91-8 Molecular Formula: Ru(NH3)6Cl3 Formula Weight: 309.61 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: Pale yellow UN No.: 3288 MDL No.: MFCD00011478

Formula: H₁₈Cl₃N₆Ru
MW: 309.61 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00011478
CAS Number: 14282-91-8
EINECS: 238-176-8
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR