Hydrochloric acid 34 - 37%, ARISTAR® PLUS for trace metal analysis, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

10401-VWNQ09 10401-VWNQ03
CA87003-253EA 477.07 CAD
CA87003-253 87003-251
Hydrochloric acid 34 - 37%, ARISTAR® PLUS for trace metal analysis, VWR Chemicals BDH®
Hydrochloric acid

High purity acid for quantitative trace metal analysis in the parts-per-billion (ppb) level.

Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –30 °C
Density: 1.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Assay 34 - 37 %
Color max. 10 APHA
Aluminum (Al) max. 1 ppb
Antimony (Sb) max. 0.5 ppb
Arsenic (As) max. 0.5 ppb
Barium (Ba) max. 0.1 ppb
Beryllium (Be) max. 0.1 ppb
Bismuth (Bi) max. 0.1 ppb
Boron (B) max. 1 ppb
Cadmium (Cd) max. 0.1 ppb
Calcium (Ca) max. 1 ppb
Cerium (Ce) max. 0.1 ppb
Cesium (Cs) max. 0.1 ppb
Chromium (Cr) max. 0.5 ppb
Cobalt (Co) max. 0.1 ppb
Copper (Cu) max. 0.5 ppb
Dysprosium (Dy) max. 0.1 ppb
Erbium (Er) max. 0.1 ppb
Europium (Eu) max. 0.1 ppb
Gadolinium (Gd) max. 0.1 ppb
Gallium (Ga) max. 0.1 ppb
Gold (Au) max. 0.5 ppb
Hafnium (Hf) max. 0.1 ppb
Holmium (Ho) max. 0.1 ppb
Indium (In) max. 0.1 ppb
Iron (Fe) max. 1 ppb
Lanthanum (La) max. 0.1 ppb
Lead (Pb) max. 0.1 ppb
Lithium (Li) max. 0.1 ppb
Lutetium (Lu) max. 0.1 ppb
Magnesium (Mg) max. 0.5 ppb
Manganese (Mn) max. 0.1 ppb
Mercury (Hg) max. 0.1 ppb
Molybdenum (Mo) max. 0.1 ppb
Neodymium (Nd) max. 0.1 ppb
Nickel (Ni) max. 0.5 ppb
Niobium (Nb) max. 0.1 ppb
Palladium (Pd) Information Only
Platinum (Pt) Information Only
Potassium (K) max. 1 ppb
Praseodymium (Pr) max. 0.1 ppb
Rhenium (Re) max. 0.1 ppb
Rhodium (Rh) max. 0.1 ppb
Rubidium (Rb) max. 0.1 ppb
Ruthenium (Ru) max. 0.1 ppb
Samarium (Sm) max. 0.1 ppb
Scandium (Sc) max. 0.1 ppb
Selenium (Se) max. 1 ppb
Silver (Ag) max. 1 ppb
Sodium (Na) max. 1 ppb
Strontium (Sr) max. 0.1 ppb
Tantalum (Ta) Information Only
Tellurium (Te) max. 0.1 ppb
Terbium (Tb) max. 0.1 ppb
Thallium (Tl) max. 0.1 ppb
Thorium (Th) max. 0.1 ppb
Thulium (Tm) max. 0.1 ppb
Tin (Sn) max. 0.5 ppb
Titanium (Ti) max. 0.5 ppb
Tungsten (W) max. 0.1 ppb
Uranium (U) max. 0.1 ppb
Vanadium (V) max. 0.5 ppb
Ytterbium (Yb) max. 0.1 ppb
Yttrium (Y) max. 0.1 ppb
Zinc (Zn) max. 1 ppb
Zirconium (Zr) max. 0.1 ppb
Bromide (Br-) max. 10 ppm
Free Chlorine (Cl2) max. 0.5 ppm
Total Phosphorus (P) max. 0.01 ppm
Total Sulfur (S) max. 0.3 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR