Hydrochloric acid 32 - 35%, ARISTAR® ULTRA, Ultrapure for trace metal analysis, VWR Chemicals BDH®
Supplier: VWR International
|
Danger
|
CA87003-218
CA87003-216
20401-VWNF07
CA87003-218EA
2692.82
CAD
CA87003-218
CA87003-216
87003-220
Hydrochloric acid 32 - 35%, ARISTAR® ULTRA, Ultrapure for trace metal analysis, VWR Chemicals BDH®
Hydrochloric acid
Ultra high purity acid for quantitative trace metal analysis at the parts-per-trillion (ppt) level.
Formula:
HCl Boiling Pt: 110 °C (1013 hPa) Melting Pt: –30 °C Density: 1.18 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8,II |
Specification Test Results
Assay | 32 - 35 % |
Aluminum (Al) | max. 20 ppt |
Antimony (Sb) | max. 20 ppt |
Arsenic (As) | max. 50 ppt |
Barium (Ba) | max. 10 ppt |
Beryllium (Be) | max. 10 ppt |
Bismuth (Bi) | max. 10 ppt |
Boron (B) | max. 100 ppt |
Cadmium (Cd) | max. 10 ppt |
Calcium (Ca) | max. 10 ppt |
Cerium (Ce) | max. 10 ppt |
Cesium (Cs) | max. 10 ppt |
Chromium (Cr) | max. 10 ppt |
Cobalt (Co) | max. 10 ppt |
Copper (Cu) | max. 10 ppt |
Dysprosium (Dy) | max. 1 ppt |
Erbium (Er) | max. 1 ppt |
Europium (Eu) | max. 1 ppt |
Gadolinium (Gd) | max. 1 ppt |
Gallium (Ga) | max. 10 ppt |
Gold (Au) | max. 50 ppt |
Hafnium (Hf) | max. 10 ppt |
Holmium (Ho) | max. 1 ppt |
Indium (In) | max. 1 ppt |
Iron (Fe) | max. 10 ppt |
Lanthanum (La) | max. 1 ppt |
Lead (Pb) | max. 10 ppt |
Lithium (Li) | max. 10 ppt |
Lutetium (Lu) | max. 10 ppt |
Magnesium (Mg) | max. 10 ppt |
Manganese (Mn) | max. 10 ppt |
Mercury (Hg) | max. 50 ppt |
Molybdenum (Mo) | max. 10 ppt |
Neodymium (Nd) | max. 1 ppt |
Nickel (Ni) | max. 20 ppt |
Niobium (Nb) | max. 1 ppt |
Palladium (Pd) | Information Only |
Platinum (Pt) | Information Only |
Potassium (K) | max. 10 ppt |
Praseodymium (Pr) | max. 1 ppt |
Rhenium (Re) | max. 10 ppt |
Rhodium (Rh) | max. 10 ppt |
Rubidium (Rb) | max. 10 ppt |
Ruthenium (Ru) | max. 10 ppt |
Samarium (Sm) | max. 1 ppt |
Scandium (Sc) | max. 10 ppt |
Selenium (Se) | Information Only |
Silver (Ag) | max. 10 ppt |
Sodium (Na) | max. 10 ppt |
Strontium (Sr) | max. 10 ppt |
Tantalum (Ta) | Information Only |
Tellurium (Te) | max. 1 ppt |
Terbium (Tb) | max. 1 ppt |
Thallium (Tl) | max. 10 ppt |
Thorium (Th) | max. 1 ppt |
Thulium (Tm) | max. 1 ppt |
Tin (Sn) | max. 20 ppt |
Titanium (Ti) | max. 10 ppt |
Tungsten (W) | max. 10 ppt |
Uranium (U) | max. 1 ppt |
Vanadium (V) | max. 10 ppt |
Ytterbium (Yb) | max. 1 ppt |
Yttrium (Y) | max. 1 ppt |
Zinc (Zn) | max. 10 ppt |
Zirconium (Zr) | max. 10 ppt |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...