Phenyl glycidyl ether ≥99.0% (by GC)

Supplier: TCI America

G0098-25G G0098-500G
TCG0098-025GEA 39.59 CAD
TCG0098-025G TCG0098-500G
Phenyl glycidyl ether ≥99.0% (by GC)
Phenyl glycidyl ether

CAS Number: 122-60-1
MDL Number: MFCD00005133
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 245
Flash Point (°C): 125
Specific Gravity (20/20): 1.11


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,2-Epoxy-3-phenoxypropane

Formula: C₉H₁₀O₂
Boiling Pt: 245 °C
Density: 1.11 (20 °C)
Flash Pt: 125 °C
MDL Number: MFCD00005133
CAS Number: 122-60-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60c22e50a3bbeeadeb9a2f99e5aab12ab67583f0&vid=60c22e50a3bbeeadeb9a2f99e5aab12ab67583f0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR