DL-Glyceric acid 20% ∼2 mol/L water

Supplier: TCI America

Synonyms: DL-2,3-Dihydroxypropionic acid, 2,3-Dihydroxypropanoic acid

D0602-25G D0602-100G
TCD0602-025GEA 226.94 CAD
TCD0602-025G TCD0602-100G
DL-Glyceric acid 20% ∼2 mol/L water
DL-Glyceric acid

CAS Number: 473-81-4
MDL Number: MFCD00065927
Molecular Formula: C3H6O4
Molecular Weight: 106.08
Form: Clear Liquid
Color: Colorless


Other Product Names / Synonyms: DL-2,3-Dihydroxypropionic Acid

Formula: C₃H₆O₄
MW: 106.08 g/mol
MDL Number: MFCD00065927
CAS Number: 473-81-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01a7cefc67d0f55c4b3fac54995c311791a40cd5&vid=ab24700c63e16ce4b93d35181a12b929ddc5b352&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR