Guanidinium hydrochloride solution 6 M, GenAR® for biotechnology, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride

Management of Change (MOC) category = R

MACR0563-26 MACR0563-10
CAMK056326EA 59918.42 CAD
CAMK056326 CAMK056310
Guanidinium hydrochloride solution 6 M, GenAR® for biotechnology, Macron Fine Chemicals™
Guanidine hydrochloride

Clear, colorless liquid. Suitable for use in biotechnology. GMP manufactured product.

Formula: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
Melting Pt: 185
Storage Temperature: Ambient
MDL Number: MFCD00013026
CAS Number: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Appearance (clear, colorless liquid) Passes Test
Cyanide (CN) Passes Test
Heavy Metals (as Pb) ≤10 ppm
Identification (by FTIR) Passes Test
ACS - Iron (Fe) ≤3 ppm
Molarity (by titrimetry) 5.8 - 6.2
pH (1:10 dilution) 4.5 - 6.5
Ultraviolet Absorbance (1.00-cm cell vs. water) - 260 nm ≤0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR