Glutaraldehyde 25% water, Practical, J.T.Baker®

Danger

Synonyms: Glutaral, Glutardialdehyde, 1,5-Pentanedial

M.O.C. Category N

BAKRM752-09
CAJTM752-9CS 3798.78 CAD
CAJTM752-9
Glutaraldehyde 25% water, Practical, J.T.Baker®
Glutaraldehyde
Formula: C₅H₈O₂
MW: 100.12
Boiling Pt: 188
Melting Pt: -10
Density: 1.06 g/cm³ (20 °C)
Flash Pt: >71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007025
CAS Number: 111-30-8
EINECS: 203-856-5
UN: 3265
ADR: 8 III
Merck Index: 13,04485

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Identification Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR