Sodium-L(+)-glutamate monohydrate 98,5-101,5 Dry_basis FCC

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: (S)-(+)-2-Aminopentanedioic acid sodium salt monohydrate, Sodium L-Glutamate Monohydrate, L(+)-Glutamic acid sodium salt monohydrate, L(+)-2-Aminopentanedioic acid sodium salt monohydrate, (S)-(+)-Glutamic acid sodium salt monohydrate, L(+)-Glutamic acid monosodium salt monohydrate, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid sodium salt monohydrate, (S)-(+)-Sodium glutamate monohydrate, L(+)-α-Glutamic acid sodium salt monohydrate, L(+)-1-Aminopropane-1,3-dicarboxylic acid sodium salt monohydrate, (S)-(+)-α-Glutamic acid sodium salt monohydrate

M.O.C. Category RL

BAKRM746-07 BAKRM746-09
CAJTM746-7EA 292.98 CAD
CAJTM746-7 CAJTM746-08 CAJTM746-09
Sodium-L(+)-glutamate monohydrate 98,5-101,5 Dry_basis FCC
Sodium-L(+)-glutamate monohydrate
Formula: C₅H₈NNaO₄·H₂O
MW: 187.13 g/mol
Melting Pt: 225…240 °C
Flash Pt: >225 °C
Storage Temperature: Ambient
MDL Number: MFCD00150138
CAS Number: 6106-04-3
EINECS: 240-313-1
Merck Index: 13,06278

Order Now

Specification Test Results

Food GMP Manufactured Product
Meets F.C.C. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
FCC - Identification Passes Test
FCC - Assay (dried basis) 98.5 - 101.5 %
FCC - Chloride (Cl) ≤0.2 %
FCC - Lead (Pb) ≤5 mg/kg
FCC - Clarity and Color of Solution Passes Test
FCC - Loss on Drying ≤0.5 %
FCC - Specific Rotation [Å]²ºD (+) 24.5 - 25.5 Degree


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR