Glycine ≥99.5% (dried basis), Ultrapure Bioreagent for Electrophoresis, Liquid Chromatography and Molecular Biology, for LC-MS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

Management of Change (MOC) category = N

BAKR4059-02 BAKR4059-07 BAKR4059-06
CAJT4059-0EA 533.76 CAD
CAJT4059-0 CAJT4059-2 CAJT4059-7 CAJT4059-6
Glycine ≥99.5% (dried basis), Ultrapure Bioreagent for Electrophoresis, Liquid Chromatography and Molecular Biology, for LC-MS, J.T.Baker®
Glycine

For electrophoresis, liquid chromatography, and molecular biology. Lot analysis on label.

Formula: NH₂CH₂COOH
MW: 75.07 g/mol
Boiling Pt: 181
Melting Pt: 83
Density: 1.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS Number: 56-40-6
EINECS: 200-272-2
Merck Index: 13,04504
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (C₂H₅NO₂) (dried basis, by non-aqueous acid- base titration) ≥99.5 %
Homogeneity by TLC Passes Test
Residue after Ignition ≤0.10 %
Loss on Drying at 105°C ≤0.2 %
Readily Carbonizable Substances Passes Test
Absorbance of a 1M Solution - 280 nm ≤0.100
RNase Activity (None Detected) Passes Test
DNase Activity (None Detected) Passes Test
Protease Activity (None Detected) Passes Test
Arsenic (As) ≤3 ppm
ACS - Heavy Metals (as Pb) ≤5 ppm
Iron (Fe) ≤0.003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR