Glycerine 99.0-101.0% (by anhydrous basis) USP, Multi-Compendial
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms:
1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol
Management of Change (MOC) category = R
BAKR2140-R
2140-01
2140-03
CAJT2140-REA
32258.92
CAD
CAJT2140-R
CAJT2140-1
CAJT2140-3
JT2140-1
JT2140-3
Glycerine 99.0-101.0% (by anhydrous basis) USP, Multi-Compendial
Glycerine
Ideal for biotechnology, cell biology and molecular biology discovery, and biopharmaceutical production.
- Food GMP manufactured product
Formula:
HOCH₂CH(OH)CH₂OH MW: 92.09 g/mol Boiling Pt: 290 Melting Pt: 18.6 Density: 1.26 g/cm³ (20 °C) Flash Pt: 177 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004722 CAS Number: 56-81-5 EINECS: 200-289-5 Merck Index: 13,04497 |
|
Specification Test Results
Meets E.P. Chemical Specifications | |
Meets U.S.P Requirements | |
Food GMP Manufactured Product | |
Meets B.P. Chemical Specifications | |
Meets F.C.C. Requirements | |
GMP Manufactured Product | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Food Chemical | |
Bulk Pharmaceutical Chemical | |
USP - Identification A | Passes Test |
USP - Identification B - Diethylene Glycol | ≤0.10 % |
USP - Identification B - Ethylene Glycol | ≤0.10 % |
USP - Identification C | Passes Test |
USP - Fatty Acids and Esters | Passes Test |
USP - Specific Gravity at 25°/25°C | ≥1.249 |
USP - Color | Passes Test |
USP - Chloride (Cl) | ≤0.001 % |
USP - Sulfate (SO₄) | ≤0.002 % |
USP - Chlorinated Compounds (as Cl) | ≤0.003 % |
USP - Assay (calculated on anhydrous basis) | 99.0 - 101.0 % |
USP - Water (by KF) (H₂O) | ≤5.0 % |
USP - Residue on Ignition | ≤0.01 % |
USP - Related Compounds: Individual Impurity | ≤0.1 % |
USP - Related Compo unds: Total Impurities | ≤1.0 % |
FCC - Identification A | Passes Test |
FCC - Identification B (FCC) | Passes Test |
FCC - Specific Gravity (FCC) | ≥1.259 |
FCC - Color | Passes Test |
FCC - Residue on Ignition | ≤0.01 % |
FCC - Chlorinated Compounds (as Cl) | ≤0.003 % |
FCC - Fatty Acids and Esters(as butyric acid) | Passes Test |
FCC - Readily Carbonizable Substances | Passes Test |
FCC - Assay (as is) (FCC) | 99.0 - 101.0 % |
FCC - Water (by KF) (H₂O) (FCC) | ≤1.0 % |
FCC - Lead (Pb) | ≤1 mg/kg |
EP/BP - Appearance of Solution | Passes Test |
EP/BP - Acidity or Alkalinity | Passes Test |
EP/BP - Refractive Index, n²ºD | 1.470 - 1.475 |
EP/BP - Aldehydes | ≤10 ppm |
EP/BP - Ester | Passes Test |
EP/BP - Halogenated Compounds | ≤35 ppm |
EP/BP - Sugars | Passes Test |
EP/BP - Chloride (Cl) | ≤10 ppm |
EP/BP - Water (H₂O) | ≤2.0 % |
EP/BP - Ash (sulfated) | ≤0.01 % |
EP/BP - Impurity A and Related Substances - Impurity A | ≤0.1 % |
EP/BP - Impurity A and Related Substances - Other Impurities with R^T
≤0.1 % |
|
EP/BP - Impurity A and Related Substances - Total Impurities R^T>Glycerol | ≤0.5 % |
EP - Assay (anhydrous basis) | 98.0 - 101.0 % |
EP/BP - Identification A | Passes Test |
EP/BP - Identification B | Passes Test |
Bacterial Endotoxin (EU/mg) | ≤0.05 |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...