PYREX® Griffin Beakers, Graduated
Supplier: Corning
1000-10 1000-20 1000-30 1000-50 1000-100 1000-150 1000-250 1000-400 1000-600 1000-800 1000-1L 1000-2L 1000-3L 1000-4L 1000-PACK 1000-100 1000-1L 1000-250 1000-400 1000-4L 1000-50 1000-600
13912-069CS 440.96 CAD
13912-069 13912-091 13912-127 13912-149 13912-160 13912-182 13912-207 13912-229 13912-240 13912-262 13912-284 13912-320 89090-522 89090-524 89091-542 470211-310 470211-314 470211-320 470211-326 470211-328 470211-330 470211-332
PYREX® Griffin Beakers, Graduated
Beakers
PYREX® Griffin Beakers are made of PYREX® borosilicate glass.
  • Very good chemical resistance
  • Thermal shock resistance
PYREX® Griffin Beakers are manufactured with uniform wall thickness to offer optimum balance between thermal shock resistance and mechanical strength.
Beakers come in sizes 250 ml and larger have a double graduated metric scale to indicate approximate content. The 10 ml size is not graduated. All sizes have an extra large marking spot.
Certifications: Meet ASTM E960 specifications.
Ordering information: The variety packMore Product Information