DURAN® Storage/Media Bottles, Glass, with Caps, SI Analytics
Supplier: DWK Life Sciences (KIMBLE)
218017353 218011753 218015455
29110-904EA 468.09 CAD
29110-904 87006-002 29110-902 87006-000
DURAN® Storage/Media Bottles, Glass, with Caps, SI Analytics
Bottles Cell Culture Bottles
Ideal for storage, mixing, and sampling.
  • Resistant to chemical attack and thermal shock
  • All components are autoclavable
These borosilicate glass bottles feature white graduation marks in milliliter increments and a marking spot.
Delivery information: Supplied with blue polypropylene screw caps and pouring rings.
Order Now
SPECIFICATIONS
Environmentally Preferable