Graduated Beakers with Handles, Stainless Steel, Polar Ware
Supplier: Polar Ware
T1062 T1063 T1064
13983-023CS 4087.84 CAD
13983-023 13983-067 13983-089
Graduated Beakers with Handles, Stainless Steel, Polar Ware
Beakers
Made of type 18-8 stainless steel and graduated in both ounces and mL
Tapered shape with spout and handle.