Glycerine ≥95%, Supelco®

Supplier: MilliporeSigma

Synonyms: Propane-1,2,3-triolGlycerol1,2,3-Propanetriol

GX0190-5 GX0190-6 GX0190-5 GX0190-6
CAGX0190-6CS 2914.9 CAD
CAGX0190-6 EM-GX0190-5 EM-GX0190-6 CAGX0190-5
Glycerine ≥95%, Supelco®
Glycerine
Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 290 °C (1013 hPa)
Melting Pt: 18.6 °C
Density: 1.26 g/cm³ (20 °C)
Flash Pt: 177 °C
MDL Number: MFCD00004722
CAS Number: 56-81-5
Merck Index: 13,04497
https://ca.vwr.com/stibo/web/std.lang.all/47/74/734774.jpg

Order Now

Specification Test Results

Assay 95% min
Acrolein, glucose, and ammonium compounds To pass test
Chloride (Cl) 0.001% max
Chlorinated compounds (as Cl) 0.003% max
Color To pass test
Description To pass test
Fatty acids and esters To pass test
Heavy metals (as Pb) 5ppm max
Identification To pass test
Readily carbonizable substances To pass test
Residue after ignition 0.01% max
Specific Gravity at 25/25°C 1.249 min
Sulfate (SO4) 0.002% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR