Glycerine ≥99.5%, GR ACS, Supelco®

Supplier: MilliporeSigma

GX0185-3 GX0185-5 GX0185-6
CAGX0185-3EA 3104 CAD
CAGX0185-3 CAGX0185-2 CAGX0185-5 CAGX0185-6
Glycerine ≥99.5%, GR ACS, Supelco®
Glycerine

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: HOCH₂CH(OH)CH₂OH
Boiling Pt: 290 °C (1013 hPa)
Melting Pt: 18.6 °C
Density: 1.26 g/cm³ (20 °C)
Flash Pt: 177 °C
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.5% min
Acrolein and glucose To pass test
Chlorinated compounds (as Cl) 0.003% max
Color (APHA) 10 max
Fatty acid esters (as butyric acid) 0.05% max
Form Clear liquid
Heavy metals (as Pb) 2ppm max
Infrared spectrum Conforms to standard
Neutrality To pass test
Residue after ignition 0.005% max
Substances darkened by sulfuric acid To pass test
Sulfate (SO4) 0.001% max
Water (H2O) 0.5% max

SPECIFICATIONS

Environmentally Preferable Safer Human and Environmental Health
Environmental certification EPA Safer chemical solvent (green circle rating)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR