GraLab® Darkroom Timer, Dimco-Gray
Supplier: Dimco-Gray
GraLab®
7-300-160R
62373-007CS 7005.58 CAD
62373-007
GraLab® Darkroom Timer, Dimco-Gray
Timers Mechanical Timers
Ideal for photographic, graphic arts, and printing darkroom use.
  • Provides 60 minutes of timing by minutes and seconds in one-second increments
  • Buzzer alarm with variable volume control signals end of time period
Black face has a single circle of interval time numerals. Hands and numerals are phosphorescent. With two three-wire outlet receptacles. Chemical- and moisture-resistant case is made from a blend of hybrid plastics.
Certifications: UL listed.