Guanidinium thiocyanate, OmniPur® for biotechnology, Millipore®

Supplier: MilliporeSigma
Warning

Synonyms: Guanidinium sulfocyanide, Guanidinium sulphocyanide, GTC, Guanidine Thiocyanate, Guanidinium rhodanide

5120-250GM 5140-1KG
CA-EM5120EA 351.67 CAD
CA-EM5120 CA-EM5140 CAEM5150
Guanidinium thiocyanate, OmniPur® for biotechnology, Millipore®
Guanidinium thiocyanate

For molecular biology. DNase-, RNase-, and protease-free.

Formula: CH₅N₃·CHNS
Melting Pt: 118 °C
Density: 1.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013027
CAS Number: 593-84-0
EINECS: 209-812-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR