Guanidinium hydrochloride, OmniPur®, Calbiochem®, Millipore®

Supplier: MilliporeSigma
Warning

Synonyms: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride

5010-1KG 5020-10KG 5030-5KG
CA-EM5010EA 966.25 CAD
CA-EM5010 CA5020 CAEM5030
Guanidinium hydrochloride, OmniPur®, Calbiochem®, Millipore®
Guanidine hydrochloride

For molecular biology. DNase-, RNase-, protease-, and phosphatase-free.

Formula: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
Melting Pt: 185 (lit.)
Density: 1.3 g/cm3 (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00013026
CAS Number: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR