Glycine ≥99.0%, OmniPur®, Millipore®

Supplier: MilliporeSigma

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

4810-500GM 4840-5KG 4860-25KG
CA-EM4810EA 276.3 CAD
CA-EM4810 CA-EM4840 CAEM4860
Glycine ≥99.0%, OmniPur®, Millipore®
Glycine

For molecular biology. DNase-, RNase-, protease-, and phosphatase-free.

Formula: NH₂CH₂COOH
MW: 75.07 g/mol
Boiling Pt: 181
Melting Pt: 240 °C (dec., lit.)
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS Number: 56-40-6
EINECS: 200-272-2
Merck Index: 13,04504
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR