Glycerine ≥99.0%, OmniPur®, Millipore®

Supplier: MilliporeSigma

Synonyms: 1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol

4750-1L 4760-4L
CA-EM4750EA 550.48 CAD
CA-EM4750 CA4760
Glycerine ≥99.0%, OmniPur®, Millipore®
Glycerine

For molecular biology. DNase-, RNase-, protease-, and phosphatse-free. Passes test for identification.

Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 290
Melting Pt: 18.6
Density: 1.26 g/cm³ (20 °C)
Flash Pt: 177 °C
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR