Glycerine ≥99.7% ACS, Laboratory Reagent, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: 1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol

BDH1172-19L BDH1172-4LP BDH1172-1LP
BDH1172-19LEA 6092.09 CAD
BDH1172-19L BDH1172-4LP BDH1172-1LP
Glycerine ≥99.7% ACS, Laboratory Reagent, VWR Chemicals BDH®
Glycerine

Clear liquid. Suitable for organic synthesis, cleaning, and prep scale synthesis.

Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 290
Melting Pt: 18.6
Density: 1.26 g/cm³ (20 °C)
Flash Pt: 177 °C
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear
Identification Passes test
Water (K.F.) Less than 0.3%
Residue after Ignition Less than 0.01%
Assay (calc from Specific Gravity, use % Glycerine test) Greater than 99.7%
Color (APHA) Less than 20
Specific Gravity (25ºC/25ºC) 1.2613 or greater
Chlorides Less than 10 ppm
Sulfates Less than 20 ppm
Heavy Metals (as Pb) Less than 5 ppm
Chlorinated Compounds (as Cl) Less than 30 ppm
Fatty Acids and Esters Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR