Germanium ≥99.999% (metals basis), powder -100 mesh

Supplier: Thermo Scientific Chemicals

10190-09 10190-18
CAAA10190-09EA 303.36 CAD
CAAA10190-09 CAAA10190-18
Germanium ≥99.999% (metals basis), powder -100 mesh
Germanium

MDL: MFCD00085310


Purity: 99.999% CAS No.: 7440-56-4 Odor: Odorless MDL No.: MFCD00085310

Formula: Ge
MW: 72.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00085310
CAS Number: 7440-56-4
EINECS: 231-164-3
UN: 3089
ADR: 4.1,III

Order Now

Specification Test Results

Assay (by Metals Basis) 99.999%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR