4-Fluorophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
F0361-5G
F0361-25G
TCF0361-005GEA
58.46
CAD
TCF0361-005G
TCF0361-025G
4-Fluorophenylboronic acid (contains varying amounts of Anhydride)
4-Fluorophenylboronic acid
CAS Number: 1765-93-1
MDL Number: MFCD00039136
Molecular Formula: C6H6BFO2
Molecular Weight: 139.92
Form: Crystal
Color: White
Melting point (°C): 265
Other Product Names / Synonyms: 4-Fluorobenzeneboronic Acid
Formula:
C₆H₆BFO₂ Melting Pt: 265 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039136 CAS Number: 1765-93-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...