Triethyl-2-fluoro-2-phosphonoacetate (fluorinated horner reagent) ≥95.0%

Supplier: TCI America

F0340-5G F0340-1G
TCF0340-005GEA 124.82 CAD
TCF0340-005G TCF0340-001G
Triethyl-2-fluoro-2-phosphonoacetate (fluorinated horner reagent) ≥95.0%
Triethyl 2-fluoro-2-phosphonoacetate (fluorinated Horner reagent)

CAS Number: 2356-16-3
MDL Number: MFCD00134400
Molecular Formula: C8H16FO5P
Molecular Weight: 242.18
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 200
Flash Point (°C): 73
Specific Gravity (20/20): 1.19


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination, Other Fluorinated Group Introducing Agents
Other Product Names / Synonyms: (Diethoxyphosphinyl)fluoroacetic Acid Ethyl Ester, Ethyl (Diethoxyphosphinyl)fluoroacetate, 2-Fluoro-2-phosphonoacetic Acid Triethyl Ester

Formula: C₈H₁₆FO₅P
Boiling Pt: 200 °C
Density: 1.19 (20 °C)
Flash Pt: 73 °C
MDL Number: MFCD00134400
CAS Number: 2356-16-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5b7c796df589251157e12ac4fdb5647a637a287&vid=a5b7c796df589251157e12ac4fdb5647a637a287&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR