3-Furoic acid 98,0

Supplier: TCI America

Synonyms: Furan-3-carboxylic acid, 3-Furancarboxylic acid

F0304-5G F0304-25G
TCF0304-005GEA 142.2 CAD
TCF0304-005G TCF0304-025G
3-Furoic acid 98,0
3-Furoic acid

CAS Number: 488-93-7
MDL Number: MFCD00005350
Molecular Formula: C5H4O3
Molecular Weight: 112.08
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 122


Other Product Names / Synonyms: 3-Furoic Acid

Formula: C₅H₄O₃
MW: 112.08 g/mol
Melting Pt: 122 °C
MDL Number: MFCD00005350
CAS Number: 488-93-7
EINECS: 207-689-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8271384370ef409b54740fdeafcd76b77089bd09&vid=8271384370ef409b54740fdeafcd76b77089bd09&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR