3-(4-Fluorophenoxy)benzyl bromide ≥95.0%

Supplier: TCI America

Synonyms: 3'-Bromomethyl-4-fluorodiphenyl Ether

F0243-5G F0243-1G
TCF0243-5GEA 1194.48 CAD
TCF0243-5G TCF0243-001G
3-(4-Fluorophenoxy)benzyl bromide ≥95.0%
3-(4-Fluorophenoxy)benzyl bromide

CAS Number: 65295-58-1
MDL Number: MFCD00059915
Molecular Formula: C13H10BrFO
Molecular Weight: 281.12
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 160
Specific Gravity (20/20): 1.43


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3'-Bromomethyl-4-fluorodiphenyl Ether

Formula: C₁₃H₁₀BrFO
MW: 281.12 g/mol
Boiling Pt: 160 °C
Density: 1.43 (20 °C)
MDL Number: MFCD00059915
CAS Number: 65295-58-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ad58964acdfdda4f1ed2ba279449087e0d6139f&vid=4ad58964acdfdda4f1ed2ba279449087e0d6139f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR