Formaldehyde 10% in aqueous solution stabilised histological fixative, buffered (pH 7.0)

Supplier: Ricca Chemical
Danger

Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde

R3190000-4A R3190000-10F R3190000-1A R3190000-20F
RC31901EA 191.18 CAD
RC31901 RC319025 RC319032 RC31905
Formaldehyde 10% in aqueous solution stabilised histological fixative, buffered (pH 7.0)
Formaldehyde

Contains neutral phosphate buffer.

Formula: CH₂O
MW: 30.03 g/mol
Boiling Pt: -19.3
Melting Pt: -92
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
EINECS: 200-001-8
UN: 3334
ADR: 9
Merck Index: 13,04259

Order Now

Specification Test Results

Appearance Clear, colorless, Formaldehyde odor
pH at 25 °C (traceable to NIST SRM 186 & 191) 7.05 ± 0.05 pH at 25.0 °C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR