Formaldehyde 10% (v/v) in aqueous solution histological fixative, (non-buffered)

Supplier: Ricca Chemical
Danger

Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde

R3180000-10F R3180000-1A R3180000-20F
RC318025EA 222.78 CAD
RC318025 RC318032 RC31805
Formaldehyde 10% (v/v) in aqueous solution histological fixative, (non-buffered)
Formaldehyde
Formula: CH₂O
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
EINECS: 200-001-8
UN: 3334
ADR: 9,

Order Now

Specification Test Results

Appearance Clear, colorless, Formaldehyde odor


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR