Ferroin (1,10-phenanthrolineferrous sulfate complex) (7 mg/mL FeSO₄) in aqueous solution indicator, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Tris(1,10-phenanthroline)iron (II) sulphate, Tris(1,10-phenanthroline-N1,N10)iron sulphate, Tris(1,10-phenanthroline)iron (II) sulfate, Tris(1,10-phenanthroline-N1,N10)iron sulfate

Management of Change (MOC) category = N

H119-01 H119-03
MKH11901CS 1762 CAD
MKH11901 MKH11903
Ferroin (1,10-phenanthrolineferrous sulfate complex) (7 mg/mL FeSO₄) in aqueous solution indicator, Macron Fine Chemicals™
Ferroin (1,10-Phenanthroline-ferrous sulphate-complex)

APHA for COD. Aqueous, composed of 1.5% 1,10 phenanthroline and 7.5% ferrous sulfate.

Formula: C₃₆H₂₄FeN₆SO₄
MW: 692.54
Density: 0.9999
MDL Number: MFCD00036428
CAS Number: 14634-91-4
EINECS: 238-676-6

Order Now

Specification Test Results

For Laboratory, Research, or Manufacturing Use
Suitability Passes Test
Appearance (clear, dark red solution)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR