Florisil® 60-100 mesh for pesticide residue analysis, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Talc, Magnesium silicate, activated, Silicic acid magnesium salt, Florisil

Management of Change (MOC) category = N

MACR6877-06 MACR6877-08
CAMK687706CS 6218.85 CAD
CAMK687706 CAMK687708
Florisil® 60-100 mesh for pesticide residue analysis, Macron Fine Chemicals™
Magnesium silicate

Suitable for use in pesticide residue analysis.

Formula: MgO₃Si
Density: 2.51 g/cm³
MDL Number: MFCD00078249
CAS Number: 1343-88-0
EINECS: 215-681-1

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
GLC Interference Passes Test
On U.S. No. 60 Sieve ≤15 %
Thru U.S. No. 60 Sieve ≥75 %
On U.S. No. 100 Sieve ≥75 %
Thru U.S. No. 100 Sieve ≤10 %
Appearance (fine, white crystalline powder)
Product may contain black particles that are produced during the manufacturing process.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR