Formamide ≥99.5% (by GC), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Carbamaldehyde

Management of Change (MOC) category = N

M522-06 BAKRM522-06
CAJTM522-6CS 9734.05 CAD
JTM522-6 CAJTM522-6
Formamide ≥99.5% (by GC), BAKER ANALYZED® ACS, J.T.Baker®
Formamide

Lot analysis on label.

Formula: HCONH₂
MW: 45.04 g/mol
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: 2 °C
Density: 1.1334 g/cm³ (20 °C)
Flash Pt: 79 °C
MDL Number: MFCD00007941
CAS Number: 75-12-7
Merck Index: 13,04261
https://ca.vwr.com/stibo/web/std.lang.all/25/02/2302502.jpg

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (HCONH₂) (by GC) ≥99.5 %
Freezing Point 2.0 - 3.0 °C
Color (APHA) ≤10
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR