Florisil® 100-200 mesh

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Talc, Magnesium silicate, activated, Silicic acid magnesium salt, Florisil

BAKRM369-07 BAKRM369-08
CAJTM369-7EA 486.7 CAD
CAJTM369-7 CAJTM369-8
Florisil® 100-200 mesh
Magnesium silicate

Selective adsorbent for gas chromatography. Contains activated magnesium silicate.

Formula: MgO₃Si
Density: 2.51 Kg/L
MDL Number: MFCD00078249
CAS Number: 1343-88-0
EINECS: 215-681-1

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR