Florisil® 60-100 mesh

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Talc, Magnesium silicate, activated, Silicic acid magnesium salt, Florisil

Management of Change (MOC) category = N

BAKRM368-07 BAKRM368-08
CAJTM368-7EA 582.26 CAD
CAJTM368-7 CAJTM368-8
Florisil® 60-100 mesh
Magnesium silicate

Selective adsorbent for gas chromatography. Contains activated magnesium silicate.

Formula: MgO₃Si
MW: 100.38 g/mol
Density: 2.51 Kg/L
MDL Number: MFCD00078249
CAS Number: 1343-88-0
EINECS: 215-681-1

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Mesh - On U.S. No. 60 Sieve ≤10 %
Mesh - Thru U.S. No. 100 Sieve ≤10 %
Appearance (fine, white crystalline powder). However, due to the high temperature involved in the activation process, some small
charred particles may be visible within the product.
Florisil® - Registered Trademark of Floridin Company


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR